Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCTTGCAAGATGGAGGGCTGCA[G/T]GTGGTGGAAAAGCAGAACCTTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHGDH - phosphoglycerate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006623.3 | 373 | Missense Mutation | CAG,CAT | Q,H 29 | NP_006614.2 | |
XM_011541226.1 | 373 | Missense Mutation | CAG,CAT | Q,H 29 | XP_011539528.1 | |
XM_011541227.2 | 373 | Intron | XP_011539529.1 | |||
XM_011541228.1 | 373 | Intron | XP_011539530.1 | |||
XM_011541231.2 | 373 | UTR 5 | XP_011539533.1 |