Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCCCTGCAGTGGACATGTCCAA[C/T]GAGATCCCCGTTCACCGTCCAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606897 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYST - lysosomal trafficking regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000081.3 | 11705 | Missense Mutation | ATT,GTT | I,V 3696 | NP_000072.2 | |
NM_001301365.1 | 11705 | Missense Mutation | ATT,GTT | I,V 3696 | NP_001288294.1 | |
XM_011544031.1 | 11705 | Missense Mutation | ATT,GTT | I,V 3750 | XP_011542333.1 | |
XM_011544032.1 | 11705 | Missense Mutation | ATT,GTT | I,V 3750 | XP_011542334.1 | |
XM_011544033.2 | 11705 | Missense Mutation | ATT,GTT | I,V 3750 | XP_011542335.1 | |
XM_011544034.1 | 11705 | Missense Mutation | ATT,GTT | I,V 3704 | XP_011542336.1 | |
XM_011544035.2 | 11705 | Intron | XP_011542337.1 | |||
XM_011544036.2 | 11705 | Missense Mutation | ATT,GTT | I,V 2971 | XP_011542338.1 | |
XM_011544037.2 | 11705 | Intron | XP_011542339.1 | |||
XM_011544039.2 | 11705 | Intron | XP_011542341.1 | |||
XM_011544040.2 | 11705 | Intron | XP_011542342.1 | |||
XM_017000150.1 | 11705 | Missense Mutation | ATT,GTT | I,V 3673 | XP_016855639.1 | |
XM_017000151.1 | 11705 | Intron | XP_016855640.1 |