Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGACTGTGCACTCGTGTGTG[C/T]GTGAGAGATCTTTATTGAGCAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
THEMIS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THEMIS2 - thymocyte selection associated family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039477.2 | Intron | NP_001034566.1 | ||||
NM_001105556.2 | Intron | NP_001099026.1 | ||||
NM_001286113.1 | Intron | NP_001273042.1 | ||||
NM_001286115.1 | Intron | NP_001273044.1 | ||||
NM_004848.3 | Intron | NP_004839.2 | ||||
XM_005246041.3 | Intron | XP_005246098.1 | ||||
XM_006711050.1 | Intron | XP_006711113.1 | ||||
XM_011542445.2 | Intron | XP_011540747.1 |