Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAAATCAGATACTTTAAAAATA[A/C]CAGAGAGCCTACTATTTAGATGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614638 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DESI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DESI2 - desumoylating isopeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297746.1 | Intron | NP_001284675.1 | ||||
NM_016076.4 | Intron | NP_057160.2 | ||||
XM_011544203.2 | Intron | XP_011542505.1 | ||||
XM_011544205.2 | Intron | XP_011542507.1 | ||||
XM_011544206.2 | Intron | XP_011542508.1 | ||||
XM_011544207.2 | Intron | XP_011542509.1 |