Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCCGTTGGTTATGCCGTCGTTC[C/T]TCAGGGGTACAGGTTGACTGGGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607999 | ||||||||||||||||||||
Literature Links: |
ASH1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASH1L - ASH1 like histone lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018489.2 | 3702 | Silent Mutation | GAA,GAG | E,E 2909 | NP_060959.2 | |
XM_005245337.4 | 3702 | Intron | XP_005245394.1 | |||
XM_006711450.3 | 3702 | Silent Mutation | GAA,GAG | E,E 2909 | XP_006711513.1 | |
XM_006711451.3 | 3702 | Silent Mutation | GAA,GAG | E,E 2909 | XP_006711514.1 | |
XM_006711452.3 | 3702 | Silent Mutation | GAA,GAG | E,E 2849 | XP_006711515.1 | |
XM_011509769.2 | 3702 | Intron | XP_011508071.1 | |||
XM_011509770.2 | 3702 | Silent Mutation | GAA,GAG | E,E 1189 | XP_011508072.1 | |
XM_017001784.1 | 3702 | Silent Mutation | GAA,GAG | E,E 2909 | XP_016857273.1 | |
XM_017001785.1 | 3702 | Silent Mutation | GAA,GAG | E,E 2909 | XP_016857274.1 | |
XM_017001786.1 | 3702 | Intron | XP_016857275.1 | |||
XM_017001787.1 | 3702 | Intron | XP_016857276.1 | |||
XM_017001788.1 | 3702 | Silent Mutation | GAA,GAG | E,E 1189 | XP_016857277.1 |
MIR555 - microRNA 555 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUSC1 - RUN and SH3 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |