Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCGCCTGCAATCTGGCCTGGCTG[A/G]TGGCCAAAGCCTTTGGGACAGGTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613775 | ||||||||||||||||||||
Literature Links: |
CAMSAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMSAP2 - calmodulin regulated spectrin associated protein family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297707.1 | 388 | Missense Mutation | ATG,GTG | M,V 40 | NP_001284636.1 | |
NM_001297708.1 | 388 | Missense Mutation | ATG,GTG | M,V 40 | NP_001284637.1 | |
NM_203459.2 | 388 | Missense Mutation | ATG,GTG | M,V 40 | NP_982284.1 | |
XM_005245041.3 | 388 | Missense Mutation | ATG,GTG | M,V 40 | XP_005245098.1 | |
XM_017000799.1 | 388 | Nonsense Mutation | TGA,TGG | *,W 15 | XP_016856288.1 |