Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGATCTTATGTATTAATCTGA[A/C]GGCTGCTAATTTTTCTAAGTACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM34 - RNA binding motif protein 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA14B - small nucleolar RNA, H/ACA box 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOMM20 - translocase of outer mitochondrial membrane 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014765.2 | Intron | NP_055580.1 |