Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTGGCCAGCACGTAGCCCAGCT[G/T]CTGGAAGAAGTGGGCTCGGCTCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601365 | ||||||||||||||||||||
Literature Links: |
DVL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DVL1 - dishevelled segment polarity protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MXRA8 - matrix remodeling associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282582.1 | 1311 | Missense Mutation | AAG,CAG | K,Q 341 | NP_001269511.1 | |
NM_001282583.1 | 1311 | Missense Mutation | AAG,CAG | K,Q 332 | NP_001269512.1 | |
NM_001282584.1 | 1311 | Missense Mutation | AAG,CAG | K,Q 240 | NP_001269513.1 | |
NM_001282585.1 | 1311 | Missense Mutation | AAG,CAG | K,Q 341 | NP_001269514.1 | |
NM_032348.3 | 1311 | Missense Mutation | AAG,CAG | K,Q 341 | NP_115724.1 | |
XM_017001516.1 | 1311 | Missense Mutation | AAG,CAG | K,Q 341 | XP_016857005.1 | |
XM_017001517.1 | 1311 | Missense Mutation | AAG,CAG | K,Q 240 | XP_016857006.1 |