Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTTATTGGGAATAAATGCCAGC[A/G]CTGCACAAATTCAGAAAAGAAGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM76A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM76A - family with sequence similarity 76 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143912.1 | 464 | Missense Mutation | CAC,CGC | H,R 121 | NP_001137384.1 | |
NM_001143913.1 | 464 | Missense Mutation | CAC,CGC | H,R 121 | NP_001137385.1 | |
NM_001143914.1 | 464 | Missense Mutation | CAC,CGC | H,R 87 | NP_001137386.1 | |
NM_001143915.1 | 464 | Intron | NP_001137387.1 | |||
NM_152660.2 | 464 | Missense Mutation | CAC,CGC | H,R 87 | NP_689873.1 | |
XM_017000543.1 | 464 | Missense Mutation | CAC,CGC | H,R 87 | XP_016856032.1 |