Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCAACAAGGACAACTCTCTCAT[A/G]TCTCTTGCACAGAAGTCATACCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605541 | ||||||||||||||||||||
Literature Links: |
VAV3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VAV3 - vav guanine nucleotide exchange factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079874.1 | 1339 | Silent Mutation | GAC,GAT | D,D 243 | NP_001073343.1 | |
NM_006113.4 | 1339 | Silent Mutation | GAC,GAT | D,D 803 | NP_006104.4 | |
XM_005270360.2 | 1339 | Silent Mutation | GAC,GAT | D,D 709 | XP_005270417.1 | |
XM_005270361.1 | 1339 | Intron | XP_005270418.1 | |||
XM_017000053.1 | 1339 | Silent Mutation | GAC,GAT | D,D 779 | XP_016855542.1 | |
XM_017000054.1 | 1339 | Intron | XP_016855543.1 | |||
XM_017000055.1 | 1339 | Silent Mutation | GAC,GAT | D,D 707 | XP_016855544.1 | |
XM_017000056.1 | 1339 | Silent Mutation | GAC,GAT | D,D 363 | XP_016855545.1 |