Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCGGCTCTTGCCGATGGCCCTC[C/T]GGAGGGTCTTCTGCAGGGCTTGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3BP5L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH3BP5L - SH3 binding domain protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322462.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001309391.1 | |
NM_001322463.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001309392.1 | |
NM_001322464.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 49 | NP_001309393.1 | |
NM_030645.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 216 | NP_085148.1 | |
XM_017002404.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 74 | XP_016857893.1 | |
XM_017002405.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 74 | XP_016857894.1 | |
XM_017002406.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 74 | XP_016857895.1 | |
XM_017002407.1 | 1087 | Missense Mutation | CAG,CGG | Q,R 184 | XP_016857896.1 |