Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGAAGCCCAACAGGTAGAACTT[C/T]ATGCAGAGCTCCAGGCCATGAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UAP1 - UDP-N-acetylglucosamine pyrophosphorylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324113.1 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | NP_001311042.1 | |
NM_001324114.1 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | NP_001311043.1 | |
NM_001324115.1 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | NP_001311044.1 | |
NM_001324116.1 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | NP_001311045.1 | |
NM_001324117.1 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | NP_001311046.1 | |
NM_003115.4 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | NP_003106.3 | |
XM_011509913.2 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | XP_011508215.1 | |
XM_011509914.2 | 380 | Missense Mutation | CAT,TAT | H,Y 34 | XP_011508216.1 |