Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTACTATTAATAGATTTGCCAAC[A/G]TAAGAAAGTTGGTGACTTTAAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM163A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM163A - family with sequence similarity 163 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173509.2 | Intron | NP_775780.1 | ||||
XM_005244913.2 | Intron | XP_005244970.1 | ||||
XM_006711174.2 | Intron | XP_006711237.1 | ||||
XM_006711175.2 | Intron | XP_006711238.1 | ||||
XM_006711176.2 | Intron | XP_006711239.1 | ||||
XM_006711177.1 | Intron | XP_006711240.1 | ||||
XM_011509217.1 | Intron | XP_011507519.1 | ||||
XM_011509221.1 | Intron | XP_011507523.1 | ||||
XM_011509226.1 | Intron | XP_011507528.1 | ||||
XM_011509228.1 | Intron | XP_011507530.1 | ||||
XM_017000386.1 | Intron | XP_016855875.1 | ||||
XM_017000387.1 | Intron | XP_016855876.1 | ||||
XM_017000388.1 | Intron | XP_016855877.1 | ||||
XM_017000389.1 | Intron | XP_016855878.1 | ||||
XM_017000390.1 | Intron | XP_016855879.1 | ||||
XM_017000391.1 | Intron | XP_016855880.1 | ||||
XM_017000392.1 | Intron | XP_016855881.1 | ||||
XM_017000393.1 | Intron | XP_016855882.1 |