Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGTGAGTACCAGATTAAAAATG[C/T]GTTCTGTTGACATAACATTGGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC30A10 - solute carrier family 30 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018713.2 | 1996 | Silent Mutation | ACC,ACT | T,T 483 | NP_061183.2 | |
XM_006711437.3 | 1996 | Silent Mutation | ACC,ACT | T,T 420 | XP_006711500.2 | |
XM_017001684.1 | 1996 | Silent Mutation | ACC,ACT | T,T 324 | XP_016857173.1 |