Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACTTCCGCTGCCAGAATGGCAG[A/G]TGCATCCCCTCAAGCCTCGTGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 142461 | ||||||||||||||||||||
Literature Links: |
HSPG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPG2 - heparan sulfate proteoglycan 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LDLRAD2 - low density lipoprotein receptor class A domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013693.2 | 736 | Silent Mutation | AGA,AGG | R,R 183 | NP_001013715.2 | |
XM_005245873.4 | 736 | Silent Mutation | AGA,AGG | R,R 183 | XP_005245930.1 |