Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGCCCCACTCCTGCCTTTCTA[C/T]ACTTGTGTAGAGATATAGCCAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 134390 | ||||||||||||||||||||
Literature Links: |
F3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
F3 - coagulation factor III, tissue factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178096.1 | 1051 | Missense Mutation | ATA,GTA | I,V 224 | NP_001171567.1 | |
NM_001993.4 | 1051 | Missense Mutation | TAT,TGT | Y,C 277 | NP_001984.1 |