Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGATGGGACTCTTCAGACAATC[C/T]AAACATCCAATGGCCAATATCACCT
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||
Literature Links: |
OR2T4 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
SAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
EAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
EUR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
AFR - Not Available | |||||
OR2T4 - olfactory receptor family 2 subfamily T member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004696.1 | Intron | NP_001004696.1 |