Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCGTGCACTACGCAGGCCTCCA[C/T]GTCTGCGGCGGCTCCATCCTCAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PRSS38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRSS38 - protease, serine 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183062.2 | 249 | Silent Mutation | CAC,CAT | H,H 83 | NP_898885.1 | |
XM_011544175.2 | 249 | Missense Mutation | CAC,CAT | H,H 83 | XP_011542477.1 | |
XM_011544176.2 | 249 | Missense Mutation | CAC,CAT | H,H 83 | XP_011542478.1 |