Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACCTTCAGAGAGTCCTCGAACA[A/G]AGAAATTTCCACTAACTGAAGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605494 | ||||||||||||||||||||
Literature Links: |
EFCAB7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFCAB7 - EF-hand calcium binding domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032437.3 | 323 | Missense Mutation | AAG,GAG | K,E 25 | NP_115813.2 | |
XM_005271274.3 | 323 | Missense Mutation | AAG,GAG | K,E 25 | XP_005271331.1 | |
XM_006710976.3 | 323 | Missense Mutation | AAG,GAG | K,E 38 | XP_006711039.1 | |
XM_006710977.1 | 323 | Missense Mutation | AAG,GAG | K,E 25 | XP_006711040.1 | |
XM_011542301.2 | 323 | Missense Mutation | AAG,GAG | K,E 38 | XP_011540603.1 | |
XM_017002548.1 | 323 | Missense Mutation | AAG,GAG | K,E 38 | XP_016858037.1 |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |