Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCGCTAAGAGTGTTTGATCTT[C/T]CTCTGACTTGGTACTATTCTCTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616773 | ||||||||||||||||||||
Literature Links: |
MIGA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIGA1 - mitoguardin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270384.1 | 168 | Missense Mutation | CCT,TCT | P,S 56 | NP_001257313.1 | |
NM_198549.3 | 168 | Missense Mutation | CCT,TCT | P,S 56 | NP_940951.1 | |
XM_005270846.4 | 168 | Missense Mutation | CCT,TCT | P,S 56 | XP_005270903.1 | |
XM_005270847.4 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_005270904.1 | |
XM_005270848.4 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_005270905.1 | |
XM_011541422.2 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_011539724.1 | |
XM_017001227.1 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_016856716.1 | |
XM_017001228.1 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_016856717.1 | |
XM_017001229.1 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_016856718.1 | |
XM_017001230.1 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_016856719.1 | |
XM_017001231.1 | 168 | Missense Mutation | CCT,TCT | P,S 56 | XP_016856720.1 | |
XM_017001232.1 | 168 | Missense Mutation | CCT,TCT | P,S 24 | XP_016856721.1 |