Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGCGGAAGCCCTTTACACTACG[A/G]TGTTTCCGGCTTCAAGATGGTCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601902 MIM: 617031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ORC1 - origin recognition complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190818.1 | 64 | Intron | NP_001177747.1 | |||
NM_001190819.1 | 64 | Intron | NP_001177748.1 | |||
NM_004153.3 | 64 | Intron | NP_004144.2 | |||
XM_011541527.2 | 64 | Intron | XP_011539829.1 | |||
XM_017001388.1 | 64 | Intron | XP_016856877.1 | |||
XM_017001389.1 | 64 | Intron | XP_016856878.1 |
PRPF38A - pre-mRNA processing factor 38A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032864.3 | 64 | UTR 5 | NP_116253.2 | |||
XM_011542315.2 | 64 | UTR 5 | XP_011540617.1 |