Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGATCCACCCCCAAAACCGAC[A/G]TCTTGAGGCTGGTGAGCCCCCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602746 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100996583 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100996583 - uncharacterized LOC100996583 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC115110 - uncharacterized LOC115110 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF14 - TNF receptor superfamily member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297605.1 | 358 | Missense Mutation | ATC,GTC | I,V 20 | NP_001284534.1 | |
NM_003820.3 | 358 | Missense Mutation | ATC,GTC | I,V 20 | NP_003811.2 | |
XM_006711019.2 | 358 | Missense Mutation | ATC,GTC | I,V 20 | XP_006711082.1 | |
XM_011542383.2 | 358 | Intron | XP_011540685.1 | |||
XM_017002718.1 | 358 | Intron | XP_016858207.1 | |||
XM_017002719.1 | 358 | Intron | XP_016858208.1 | |||
XM_017002720.1 | 358 | UTR 5 | XP_016858209.1 | |||
XM_017002721.1 | 358 | UTR 5 | XP_016858210.1 |