Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTCAGCAACTTCTCAATGAGGA[G/T]AATTTACGGAAGCAGGAGGAGTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612316 MIM: 612317 | ||||||||||||||||||||
Literature Links: |
ATAD3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAD3A - ATPase family, AAA domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170535.1 | 732 | Missense Mutation | GAG,GAT | E,D 154 | NP_001164006.1 | |
NM_001170536.1 | 732 | Missense Mutation | GAG,GAT | E,D 75 | NP_001164007.1 | |
NM_018188.3 | 732 | Missense Mutation | GAG,GAT | E,D 202 | NP_060658.3 | |
XM_017001628.1 | 732 | Missense Mutation | GAG,GAT | E,D 154 | XP_016857117.1 |
ATAD3B - ATPase family, AAA domain containing 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |