Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCGGAGGAACCCCTTCTCCCC[C/T]TAGGCTTGCGTCTCTGTTCCCTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBPHL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYBPHL - myosin binding protein H like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010985.2 | Intron | NP_001010985.2 | ||||
NM_001265613.1 | Intron | NP_001252542.1 | ||||
XM_017001173.1 | Intron | XP_016856662.1 | ||||
XM_017001174.1 | Intron | XP_016856663.1 | ||||
XM_017001175.1 | Intron | XP_016856664.1 |