Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCGGGAGGTGATGCTGGAGAACT[A/C]TGGGAACCTGGTCTCAGTGGGTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZFP69B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP69B - ZFP69 zinc finger protein B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023070.2 | 650 | Missense Mutation | TAT,TCT | Y,S 107 | NP_075558.2 | |
XM_005271136.1 | 650 | Missense Mutation | TAT,TCT | Y,S 107 | XP_005271193.1 | |
XM_005271137.2 | 650 | Missense Mutation | TAT,TCT | Y,S 107 | XP_005271194.1 | |
XM_005271138.3 | 650 | Missense Mutation | TAT,TCT | Y,S 5 | XP_005271195.1 | |
XM_005271139.3 | 650 | Missense Mutation | TAT,TCT | Y,S 5 | XP_005271196.1 | |
XM_017002147.1 | 650 | Missense Mutation | TAT,TCT | Y,S 107 | XP_016857636.1 |