Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGGATGAGCTCCAGGTTTTCTT[C/T]GGGCCGATGCTTCGTGTCCTCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611824 MIM: 605138 MIM: 609501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL9 - mitochondrial ribosomal protein L9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300733.1 | 708 | Missense Mutation | AAA,GAA | K,E 91 | NP_001287662.1 | |
NM_031420.3 | 708 | Missense Mutation | AAA,GAA | K,E 91 | NP_113608.1 |
OAZ3 - ornithine decarboxylase antizyme 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134939.1 | 708 | UTR 5 | NP_001128411.1 | |||
NM_001301371.1 | 708 | Intron | NP_001288300.1 | |||
NM_016178.2 | 708 | Intron | NP_057262.2 |
TDRKH - tudor and KH domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |