Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATTTTCCTTTCTCTCCAGGAGT[C/T]CCGACCTCTACATGTTGTGCTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM63A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM63A - family with sequence similarity 63 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRUNE1 - prune exopolyphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303229.1 | 272 | UTR 5 | NP_001290158.1 | |||
NM_001303242.1 | 272 | Missense Mutation | TCC,TTC | S,F 15 | NP_001290171.1 | |
NM_001303243.1 | 272 | UTR 5 | NP_001290172.1 | |||
NM_021222.2 | 272 | Missense Mutation | TCC,TTC | S,F 15 | NP_067045.1 | |
XM_005245393.4 | 272 | Missense Mutation | TCC,TTC | S,F 15 | XP_005245450.1 | |
XM_011509832.2 | 272 | Intron | XP_011508134.1 | |||
XM_017001955.1 | 272 | Missense Mutation | TCC,TTC | S,F 15 | XP_016857444.1 | |
XM_017001956.1 | 272 | UTR 5 | XP_016857445.1 | |||
XM_017001957.1 | 272 | UTR 5 | XP_016857446.1 |