Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTCCATGTGGCACAAGTGGAC[A/G]TGGGCATGAGGACCTGGGCGGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373120 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373120 - uncharacterized LOC105373120 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSEN2 - presenilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000447.2 | Intron | NP_000438.2 | ||||
NM_012486.2 | Intron | NP_036618.2 | ||||
XM_005273199.3 | Intron | XP_005273256.1 | ||||
XM_017001835.1 | Intron | XP_016857324.1 | ||||
XM_017001836.1 | Intron | XP_016857325.1 |