Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAGGCCTTTCAGCAACTTACAC[A/G]ACATTGTCAAGGGCAGTTGCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLAMF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLAMF6 - SLAM family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184714.1 | 1063 | Silent Mutation | GTC,GTT | V,V 331 | NP_001171643.1 | |
NM_001184715.1 | 1063 | Silent Mutation | GTC,GTT | V,V 281 | NP_001171644.1 | |
NM_001184716.1 | 1063 | Silent Mutation | GTC,GTT | V,V 220 | NP_001171645.1 | |
NM_052931.4 | 1063 | Silent Mutation | GTC,GTT | V,V 330 | NP_443163.1 | |
XM_017000215.1 | 1063 | Intron | XP_016855704.1 | |||
XM_017000216.1 | 1063 | Silent Mutation | GTC,GTT | V,V 282 | XP_016855705.1 | |
XM_017000217.1 | 1063 | Silent Mutation | GTC,GTT | V,V 219 | XP_016855706.1 |