Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTACTCGGTGGTGGCTGGAGC[C/T]GCGGGAAGACTCCTCTTTGGCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01057 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01057 - long intergenic non-protein coding RNA 1057 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC44A3 - solute carrier family 44 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114106.2 | 173 | Silent Mutation | GCC,GCT | A,A 57 | NP_001107578.1 | |
NM_001258340.1 | 173 | Silent Mutation | GCC,GCT | A,A 57 | NP_001245269.1 | |
NM_001258341.1 | 173 | Silent Mutation | GCC,GCT | A,A 57 | NP_001245270.1 | |
NM_001258342.1 | 173 | Silent Mutation | GCC,GCT | A,A 21 | NP_001245271.1 | |
NM_001258343.1 | 173 | Silent Mutation | GCC,GCT | A,A 21 | NP_001245272.1 | |
NM_001301079.1 | 173 | Silent Mutation | GCC,GCT | A,A 9 | NP_001288008.1 | |
NM_152369.4 | 173 | Silent Mutation | GCC,GCT | A,A 9 | NP_689582.2 | |
XM_005270440.2 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_005270497.1 | |
XM_005270441.2 | 173 | Silent Mutation | GCC,GCT | A,A 9 | XP_005270498.1 | |
XM_005270442.1 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_005270499.1 | |
XM_011540632.2 | 173 | UTR 5 | XP_011538934.1 | |||
XM_011540633.2 | 173 | Intron | XP_011538935.1 | |||
XM_011540635.2 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_011538937.1 | |
XM_011540636.2 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_011538938.1 | |
XM_017000262.1 | 173 | Silent Mutation | GCC,GCT | A,A 21 | XP_016855751.1 | |
XM_017000263.1 | 173 | Silent Mutation | GCC,GCT | A,A 9 | XP_016855752.1 | |
XM_017000264.1 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_016855753.1 | |
XM_017000265.1 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_016855754.1 | |
XM_017000266.1 | 173 | Silent Mutation | GCC,GCT | A,A 57 | XP_016855755.1 |