Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGATCTTACCTTAAGGACTCGG[A/G]TCAAATGCAGCGTTTGTAAAAACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612309 MIM: 173610 | ||||||||||||||||||||
Literature Links: |
F5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
F5 - coagulation factor V | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SELP - selectin P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003005.3 | 2423 | Silent Mutation | GAC,GAT | D,D 827 | NP_002996.2 | |
XM_005245435.1 | 2423 | Silent Mutation | GAC,GAT | D,D 827 | XP_005245492.1 | |
XM_005245436.3 | 2423 | Silent Mutation | GAC,GAT | D,D 827 | XP_005245493.1 | |
XM_005245438.1 | 2423 | Silent Mutation | GAC,GAT | D,D 787 | XP_005245495.1 | |
XM_005245439.1 | 2423 | Silent Mutation | GAC,GAT | D,D 787 | XP_005245496.1 | |
XM_005245440.1 | 2423 | Silent Mutation | GAC,GAT | D,D 765 | XP_005245497.1 |