Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTGTGGCGACGCCTGTGAC[A/G]CTTCTGTTTTTCAGAACATCTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616476 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGBL4 - ATP/GTP binding protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323573.1 | 1293 | Intron | NP_001310502.1 | |||
NM_001323574.1 | 1293 | Intron | NP_001310503.1 | |||
NM_001323575.1 | 1293 | Intron | NP_001310504.1 | |||
NM_032785.3 | 1293 | Intron | NP_116174.3 | |||
XM_011542308.2 | 1293 | Intron | XP_011540610.1 | |||
XM_011542310.2 | 1293 | Intron | XP_011540612.1 | |||
XM_017002595.1 | 1293 | Intron | XP_016858084.1 | |||
XM_017002596.1 | 1293 | Intron | XP_016858085.1 | |||
XM_017002597.1 | 1293 | Intron | XP_016858086.1 | |||
XM_017002598.1 | 1293 | Intron | XP_016858087.1 |
BEND5 - BEN domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302082.1 | 1293 | Silent Mutation | AGC,AGT | S,S 173 | NP_001289011.1 | |
NM_024603.3 | 1293 | Silent Mutation | AGC,AGT | S,S 342 | NP_078879.2 | |
XM_011542141.2 | 1293 | Silent Mutation | AGC,AGT | S,S 257 | XP_011540443.1 | |
XM_011542142.2 | 1293 | Silent Mutation | AGC,AGT | S,S 173 | XP_011540444.1 | |
XM_017002331.1 | 1293 | Silent Mutation | AGC,AGT | S,S 288 | XP_016857820.1 | |
XM_017002332.1 | 1293 | Silent Mutation | AGC,AGT | S,S 257 | XP_016857821.1 | |
XM_017002333.1 | 1293 | Intron | XP_016857822.1 | |||
XM_017002334.1 | 1293 | Silent Mutation | AGC,AGT | S,S 173 | XP_016857823.1 |