Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCTCCATGTCCACACCCAGGAAC[C/T]TGGCATTTACTTCAAACTTTCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IQGAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQGAP3 - IQ motif containing GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178229.4 | 4803 | Missense Mutation | AAG,AGG | K,R 1580 | NP_839943.2 | |
XM_011509198.2 | 4803 | Missense Mutation | AAG,AGG | K,R 1585 | XP_011507500.1 | |
XM_011509200.2 | 4803 | Missense Mutation | AAG,AGG | K,R 1138 | XP_011507502.1 | |
XM_011509201.2 | 4803 | Missense Mutation | AAG,AGG | K,R 1084 | XP_011507503.1 | |
XM_017000317.1 | 4803 | Missense Mutation | AAG,AGG | K,R 1584 | XP_016855806.1 | |
XM_017000318.1 | 4803 | Missense Mutation | AAG,AGG | K,R 1446 | XP_016855807.1 |