Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGTCCGAACACCAGCTGGCAT[A/T]GGGAAGAGCATAATCCTTCAAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615653 | ||||||||||||||||||||
Literature Links: |
LOC100132111 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100132111 - uncharacterized LOC100132111 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THEM5 - thioesterase superfamily member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182578.3 | 307 | Missense Mutation | AAT,TAT | N,Y 59 | NP_872384.1 | |
XM_011509421.1 | 307 | Missense Mutation | AAT,TAT | N,Y 59 | XP_011507723.1 |