Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCATGCCGTGTGTGTGCTGGG[C/T]ACCTTGACTCAGCTTGACATCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605347 | ||||||||||||||||||||
Literature Links: |
PADI4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PADI4 - peptidyl arginine deiminase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012387.2 | 94 | Silent Mutation | GGC,GGT | G,G 22 | NP_036519.2 | |
XM_011541150.1 | 94 | Silent Mutation | GGC,GGT | G,G 22 | XP_011539452.1 | |
XM_011541151.1 | 94 | Silent Mutation | GGC,GGT | G,G 22 | XP_011539453.1 | |
XM_011541152.1 | 94 | Intron | XP_011539454.1 | |||
XM_011541153.1 | 94 | Silent Mutation | GGC,GGT | G,G 22 | XP_011539455.1 | |
XM_011541154.2 | 94 | Silent Mutation | GGC,GGT | G,G 22 | XP_011539456.1 | |
XM_011541155.1 | 94 | Silent Mutation | GGC,GGT | G,G 22 | XP_011539457.1 | |
XM_011541156.1 | 94 | Silent Mutation | GGC,GGT | G,G 22 | XP_011539458.1 | |
XM_011541157.1 | 94 | Intron | XP_011539459.1 |