Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCACCCACCATCTCATTGTAGGT[A/G]GGATTGCAGGTTTTCCGGGCCACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602838 | ||||||||||||||||||||
Literature Links: |
PIK3C2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIK3C2B - phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002646.3 | 4126 | Silent Mutation | CCC,CCT | P,P 1567 | NP_002637.3 | |
XM_005245257.2 | 4126 | Silent Mutation | CCC,CCT | P,P 1567 | XP_005245314.1 | |
XM_005245258.3 | 4126 | Silent Mutation | CCC,CCT | P,P 1540 | XP_005245315.1 | |
XM_011509630.2 | 4126 | Silent Mutation | CCC,CCT | P,P 1567 | XP_011507932.1 | |
XM_011509631.2 | 4126 | Silent Mutation | CCC,CCT | P,P 1567 | XP_011507933.1 | |
XM_011509633.2 | 4126 | Intron | XP_011507935.1 | |||
XM_017001473.1 | 4126 | Silent Mutation | CCC,CCT | P,P 1567 | XP_016856962.1 | |
XM_017001474.1 | 4126 | Silent Mutation | CCC,CCT | P,P 1567 | XP_016856963.1 | |
XM_017001475.1 | 4126 | Silent Mutation | CCC,CCT | P,P 1003 | XP_016856964.1 |