Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAATGATTTATTCATTCTCCTGG[C/T]ACGCACAGAGAAGACAGGCTGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605434 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf216 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf216 - chromosome 1 open reading frame 216 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLSPN - claspin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190481.1 | Intron | NP_001177410.1 | ||||
NM_022111.3 | Intron | NP_071394.2 | ||||
XM_011541938.2 | Intron | XP_011540240.1 | ||||
XM_011541939.2 | Intron | XP_011540241.1 | ||||
XM_011541940.2 | Intron | XP_011540242.1 | ||||
XM_011541941.2 | Intron | XP_011540243.1 |