Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTAGGTGCCCATCCAGGTCCTC[A/G]CTGTCGCTGAACAGCTCCCCGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613911 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF496 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF496 - zinc finger protein 496 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005273328.3 | 3297 | Silent Mutation | AGC,AGT | S,S 483 | XP_005273385.1 | |
XM_005273330.3 | 3297 | Silent Mutation | AGC,AGT | S,S 447 | XP_005273387.1 | |
XM_006711838.2 | 3297 | Intron | XP_006711901.1 | |||
XM_011544310.1 | 3297 | Silent Mutation | AGC,AGT | S,S 514 | XP_011542612.1 | |
XM_011544312.1 | 3297 | Silent Mutation | AGC,AGT | S,S 478 | XP_011542614.1 | |
XM_011544314.2 | 3297 | Intron | XP_011542616.1 | |||
XM_011544315.2 | 3297 | Intron | XP_011542617.1 | |||
XM_011544316.1 | 3297 | Intron | XP_011542618.1 | |||
XM_017002586.1 | 3297 | Silent Mutation | AGC,AGT | S,S 545 | XP_016858075.1 | |
XM_017002587.1 | 3297 | Silent Mutation | AGC,AGT | S,S 525 | XP_016858076.1 | |
XM_017002588.1 | 3297 | Silent Mutation | AGC,AGT | S,S 514 | XP_016858077.1 | |
XM_017002589.1 | 3297 | Silent Mutation | AGC,AGT | S,S 509 | XP_016858078.1 | |
XM_017002590.1 | 3297 | Silent Mutation | AGC,AGT | S,S 478 | XP_016858079.1 | |
XM_017002591.1 | 3297 | Silent Mutation | AGC,AGT | S,S 427 | XP_016858080.1 | |
XM_017002592.1 | 3297 | Intron | XP_016858081.1 | |||
XM_017002593.1 | 3297 | Intron | XP_016858082.1 | |||
XM_017002594.1 | 3297 | Silent Mutation | AGC,AGT | S,S 250 | XP_016858083.1 |