Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCGAGACATACCGCTACCTGAC[A/G]GACCTGTTCACCACGCTGGTGGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606644 MIM: 600932 | ||||||||||||||||||||
Literature Links: |
IGSF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF8 - immunoglobulin superfamily member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNJ9 - potassium voltage-gated channel subfamily J member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004983.2 | 380 | Silent Mutation | ACA,ACG | T,T 46 | NP_004974.2 | |
XM_017001247.1 | 380 | Silent Mutation | ACA,ACG | T,T 103 | XP_016856736.1 |