Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTGAAACACGAACTAATCTTG[A/T]GGCTGATAAATGAATTTTGAAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPL - centromere protein L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127181.2 | 1507 | Missense Mutation | ACA,TCA | T,S 348 | NP_001120653.1 | |
NM_001171182.1 | 1507 | Missense Mutation | ACA,TCA | T,S 302 | NP_001164653.1 | |
NM_033319.3 | 1507 | Missense Mutation | ACA,TCA | T,S 302 | NP_201576.1 |