Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATATCATTAGCTTTAAAAGAGA[G/T]GAGCACCTTCAAGACAATGCTGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604227 MIM: 605667 | ||||||||||||||||||||
Literature Links: |
ARPC5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARPC5 - actin related protein 2/3 complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270439.1 | 532 | Missense Mutation | ATC,CTC | I,L 87 | NP_001257368.1 | |
NM_005717.3 | 532 | Missense Mutation | ATC,CTC | I,L 84 | NP_005708.1 |
RGL1 - ral guanine nucleotide dissociation stimulator like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |