Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCCTCTGCAATGTCACCGCCT[C/T]TGTGTCCCCTCCTTCTCCTGGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610278 | ||||||||||||||||||||
Literature Links: |
PEAR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEAR1 - platelet endothelial aggregation receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080471.1 | 1332 | Silent Mutation | CTG,TTG | L,L 5 | NP_001073940.1 | |
XM_005245141.3 | 1332 | Silent Mutation | CTG,TTG | L,L 5 | XP_005245198.1 | |
XM_011509510.2 | 1332 | Silent Mutation | CTG,TTG | L,L 5 | XP_011507812.1 | |
XM_011509511.2 | 1332 | Silent Mutation | CTG,TTG | L,L 5 | XP_011507813.1 | |
XM_011509512.2 | 1332 | UTR 5 | XP_011507814.1 | |||
XM_017001234.1 | 1332 | Silent Mutation | CTC,CTT | L,L 74 | XP_016856723.1 | |
XM_017001235.1 | 1332 | Silent Mutation | CTC,CTT | L,L 74 | XP_016856724.1 | |
XM_017001236.1 | 1332 | Silent Mutation | CTG,TTG | L,L 5 | XP_016856725.1 | |
XM_017001237.1 | 1332 | UTR 5 | XP_016856726.1 | |||
XM_017001238.1 | 1332 | UTR 5 | XP_016856727.1 | |||
XM_017001239.1 | 1332 | UTR 5 | XP_016856728.1 | |||
XM_017001240.1 | 1332 | Intron | XP_016856729.1 | |||
XM_017001241.1 | 1332 | Intron | XP_016856730.1 | |||
XM_017001242.1 | 1332 | Intron | XP_016856731.1 |