Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACGAATGTCACCCCACTTTAATT[A/C]TCACAACAGCTTCTCAAGATAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KA1 - ribosomal protein S6 kinase A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006665.1 | Intron | NP_001006666.1 | ||||
NM_002953.3 | Intron | NP_002944.2 | ||||
XM_005245966.4 | Intron | XP_005246023.1 |