Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGACTGGAAGGCAAGGGCATCT[A/G]CTGTTTGTTCCTCGTGGCCCAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606421 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELMO2 - engulfment and cell motility 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318253.1 | 3051 | UTR 3 | NP_001305182.1 | |||
NM_133171.4 | 3051 | UTR 3 | NP_573403.1 | |||
NM_182764.2 | 3051 | UTR 3 | NP_877496.1 | |||
XM_005260496.3 | 3051 | UTR 3 | XP_005260553.1 | |||
XM_005260498.3 | 3051 | UTR 3 | XP_005260555.1 | |||
XM_005260499.3 | 3051 | UTR 3 | XP_005260556.1 | |||
XM_005260500.3 | 3051 | UTR 3 | XP_005260557.1 | |||
XM_005260501.4 | 3051 | UTR 3 | XP_005260558.1 | |||
XM_006723854.3 | 3051 | UTR 3 | XP_006723917.1 | |||
XM_017028009.1 | 3051 | UTR 3 | XP_016883498.1 | |||
XM_017028010.1 | 3051 | UTR 3 | XP_016883499.1 | |||
XM_017028011.1 | 3051 | UTR 3 | XP_016883500.1 |
SLC35C2 - solute carrier family 35 member C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |