Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCGCCGGCGCCCTCGCAGGTCAA[C/G]AGCAAGGCCAAGAAGACCGTGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189965 | ||||||||||||||||||||
Literature Links: |
CEBPB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEBPB - CCAAT/enhancer binding protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285878.1 | 1231 | Missense Mutation | AAC,AAG | N,K 237 | NP_001272807.1 | |
NM_001285879.1 | 1231 | Missense Mutation | AAC,AAG | N,K 62 | NP_001272808.1 | |
NM_005194.3 | 1231 | Missense Mutation | AAC,AAG | N,K 260 | NP_005185.2 |
CEBPB-AS1 - CEBPB antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |