Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTAATTTTGGTTTTACAGGTAC[A/G]TACTTGGGATCCACACAGCACTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616075 | ||||||||||||||||||||
Literature Links: |
DEFB121 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEFB121 - defensin beta 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011878.2 | 564 | Silent Mutation | TAC,TAT | Y,Y 56 | NP_001011878.1 | |
NM_001171832.1 | 564 | Silent Mutation | TAC,TAT | Y,Y 44 | NP_001165303.1 | |
XM_005260383.3 | 564 | Silent Mutation | TAC,TAT | Y,Y 44 | XP_005260440.1 |