Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACCCATGACGCGGGCCTGCACC[A/G]TCACGCGCACACAGGTGCCAGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610702 | ||||||||||||||||||||
Literature Links: |
C20orf27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C20orf27 - chromosome 20 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039140.2 | 1087 | Missense Mutation | ACG,ATG | T,M 160 | NP_001034229.1 | |
NM_001258429.1 | 1087 | Missense Mutation | ACG,ATG | T,M 135 | NP_001245358.1 | |
NM_001258430.1 | 1087 | Missense Mutation | ACG,ATG | T,M 135 | NP_001245359.1 | |
XM_011529266.2 | 1087 | Intron | XP_011527568.1 |
HSPA12B - heat shock protein family A (Hsp70) member 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |