Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACGCGCACCTGGCCGGCAGGCAC[A/G]AGGGTCTGCTGAGCCTCGCAGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNDC11 - fibronectin type III domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HELZ2 - helicase with zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037335.2 | 8791 | Silent Mutation | CTC,CTT | L,L 2633 | NP_001032412.2 | |
NM_033405.3 | 8791 | Silent Mutation | CTC,CTT | L,L 2064 | NP_208384.3 |