Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTGCAGCAGAACACGGCTTTGG[A/G]GTCCGAAAGGTAAGCTCTCCCATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162151 | ||||||||||||||||||||
Literature Links: |
PCSK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCSK2 - proprotein convertase subtilisin/kexin type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201528.1 | 522 | Silent Mutation | GGA,GGG | G,G 37 | NP_001188457.1 | |
NM_001201529.2 | 522 | Silent Mutation | GGA,GGG | G,G 56 | NP_001188458.1 | |
NM_002594.4 | 522 | Silent Mutation | GGA,GGG | G,G 56 | NP_002585.2 |