Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAGGTGGCAAATGCTGTAGTCC[A/G]TCCTCTTGGGCAGGTGTTAGGTACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RALGAPB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RALGAPB - Ral GTPase activating protein non-catalytic beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282917.1 | 368 | Missense Mutation | CAT,CGT | H,R 39 | NP_001269846.1 | |
NM_001282918.1 | 368 | Missense Mutation | CAT,CGT | H,R 39 | NP_001269847.1 | |
NM_020336.3 | 368 | Missense Mutation | CAT,CGT | H,R 39 | NP_065069.1 | |
XM_005260462.2 | 368 | Missense Mutation | CAT,CGT | H,R 39 | XP_005260519.1 | |
XM_005260463.2 | 368 | Missense Mutation | CAT,CGT | H,R 39 | XP_005260520.1 | |
XM_005260464.2 | 368 | Missense Mutation | CAT,CGT | H,R 39 | XP_005260521.1 | |
XM_005260465.2 | 368 | Missense Mutation | CAT,CGT | H,R 39 | XP_005260522.1 | |
XM_017027966.1 | 368 | Missense Mutation | CAT,CGT | H,R 39 | XP_016883455.1 | |
XM_017027967.1 | 368 | Missense Mutation | CAT,CGT | H,R 39 | XP_016883456.1 | |
XM_017027968.1 | 368 | Intron | XP_016883457.1 | |||
XM_017027969.1 | 368 | UTR 5 | XP_016883458.1 | |||
XM_017027970.1 | 368 | Intron | XP_016883459.1 |